Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-thiophenecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. M145925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M145925G 25 g
M14595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M145925G Supplier TCI America Supplier No. M145925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23806-24-8
Molecular Formula C6H5O2S
Molecular Weight (g/mol) 141.16
MDL Number MFCD00005438
InChI Key IFLKEBSJTZGCJG-UHFFFAOYSA-M
Synonym 3-methyl-2-thiophenecarboxylic acid, 3-methyl-2-thenoic acid, 2-thiophenecarboxylic acid, 3-methyl, 2-carboxy-3-methylthiophene, 3-methyl-2-thiophene carboxylic acid, 3-methyl-2-thiophenecarboxylic, 3-methyl-thiophene-2-carboxylic acid, 3-methyl-2-thiophenecarboxylicacid, pubchem5182, acmc-1cinc
PubChem CID 90269
IUPAC Name 3-methylthiophene-2-carboxylate
SMILES CC1=C(SC=C1)C([O-])=O

Specifications

Melting Point 148°C
Quantity 25 g
Formula Weight 142.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-2-thiophenecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.