Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-(tri-n-butylstannyl)pyridine, 96%

Catalog No. AAH5154003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5154003 1 g
AAH51540MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5154003 Supplier Thermo Scientific Chemicals Supplier No. H5154003
Add to Cart

CAS: 259807-97-1 | C18H33NSn | 382.179 g/mol

3-Methyl-2-(tri-n-butylstannyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-(tri-n-butylstannyl)pyridine is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259807-97-1
Molecular Formula C18H33NSn
Molecular Weight (g/mol) 382.179
MDL Number MFCD07787404
InChI Key QWDWZQDTTVYOBK-UHFFFAOYSA-N
Synonym 3-methyl-2-tributylstannyl pyridine, 2-tributylstannyl-3-methylpyridine, pyridine, 3-methyl-2-tributylstannyl, 3-methyl-2-tri-n-butylstannyl pyridine, acmc-209gnt, 3-methyl-2-tributylstannanylpyridine, 2-tri-n-butylstannyl-3-methylpyridine
PubChem CID 16427098
IUPAC Name tributyl-(3-methylpyridin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=C(C=CC=N1)C

Specifications

Quantity 1 g
UN Number UN2788
Solubility Information Sparingly soluble in water.
Formula Weight 382.16
Percent Purity 96%
Chemical Name or Material 3-Methyl-2-(tri-n-butylstannyl)pyridine
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.