Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-3-penten-2-one, (E)+(Z), 95%, Thermo Scientific™

Catalog No. AAL1354818
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1354818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1354818 Supplier Thermo Scientific Chemicals Supplier No. L1354818

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-62-8
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00151839
InChI Key ZAMCMCQRTZKGDX-SNAWJCMRSA-N
Synonym 3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene
PubChem CID 5364579
IUPAC Name (E)-3-methylpent-3-en-2-one
SMILES CC=C(C)C(=O)C

Specifications

Melting Point -70°C
Density 0.883
Boiling Point 138°C
Flash Point 34°C (93°F)
Refractive Index 1.449
Quantity 50 g
UN Number UN1224
Formula Weight 98.15
Percent Purity 95%
Chemical Name or Material 3-Methyl-3-penten-2-one, (E)+(Z)
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 209-283-7
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.