Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-4-nitroaniline 97.0+%, TCI America™
SDP

Catalog No. M167725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M167725G 25 g
M16775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M167725G Supplier TCI America Supplier No. M167725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 611-05-2
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
InChI Key XPAYEWBTLKOEDA-UHFFFAOYSA-N
Synonym 4-nitro-m-toluidine, benzenamine, 3-methyl-4-nitro, 3-methyl-4-nitro-aniline, 3-methyl-4-nitro-phenylamine, 3-methyl-4-nitrophenylamine, 4-nitro-m-toluidin, 3-methyl4-nitroaniline, pubchem12456, 3-mehtyl-4-nitroanilin, 5-amino-2-nitrotoluene
PubChem CID 11898
IUPAC Name 3-methyl-4-nitroaniline
SMILES CC1=C(C=CC(=C1)N)[N+](=O)[O-]

Specifications

Melting Point 134°C
Quantity 25 g
UN Number 2660
Formula Weight 152.15
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-4-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.