Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-5-(trifluoromethyl)pyrazole 98.0+%, TCI America™
SDP

Catalog No. M20321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20321G 1 g
M20325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20321G Supplier TCI America Supplier No. M20321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10010-93-2
Molecular Formula C5H5F3N2
Molecular Weight (g/mol) 150.104
MDL Number MFCD00156058
InChI Key DLCHCAYDSKIFIN-UHFFFAOYSA-N
Synonym 5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole
PubChem CID 139077
IUPAC Name 5-methyl-3-(trifluoromethyl)-1H-pyrazole
SMILES CC1=CC(=NN1)C(F)(F)F

Specifications

Melting Point 89°C
Color White
Quantity 1 g
Formula Weight 150.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-5-(trifluoromethyl)pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.