Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-N-methylbenzylamine 97.0+%, TCI America™
SDP

Catalog No. M28241ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28241ML 1 mL
M28245ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28241ML Supplier TCI America Supplier No. M28241ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39180-84-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD05865181
InChI Key WCTNVGNEUDTSOZ-UHFFFAOYSA-N
Synonym 3-methyl-n-methylbenzylamine, n-methyl-1-m-tolyl methanamine, 3-methylbenzyl methylamine, methyl 3-methylphenyl methyl amine, benzenemethanamine, n,3-dimethyl, acmc-20a7fx, n,3-dimethylbenzylamine, methyl 3-methylbenzyl amine, n-methyl-3-methylbenzylamine, ksc493m5b
PubChem CID 13794465
IUPAC Name N-methyl-1-(3-methylphenyl)methanamine
SMILES CC1=CC(=CC=C1)CNC

Specifications

Color Yellow
Boiling Point 92°C
Quantity 1 mL
UN Number 2735
Formula Weight 135.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-N-methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.