Learn More
Sigma Organic Chemistry 3-Methylacetophenone | 25G | 585-74-0 | MFCD00008742
Supplier: Sigma Organic Chemistry M2660725G

3'-Methylacetophenone, 25G
About This Item:
Synonym(s): Methyl m-tolyl ketone
Linear Formula: CH3C6H4COCH3
Molecular Weight: 134.18
CAS Number: 585-74-0
MDL Number: MFCD00008742
UNSPSC Code: 12352100
Purity: 0.98
Boiling Point: 218-220 C (lit.)
Melting Point: -9 C (lit.)
Density: 0.986 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.529 (lit.)
Storage: room temp
EINECS Number: 209-561-8
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.