Learn More
AdipoGen 3-Methyladenine (250 mg)
SDP
Supplier:  AdipoGen AGCR13597M250
Encompass_Preferred
Chemical. CAS: 5142-23-4. Formula: C6H7N5. MW: 149.2. Potent cell permeable and selective inhibitor of phosphatidyl-inositol 3-kinase (PI3K). Blocks class I, class II and class III PI3Ks, including some downstream targets. Blocks class I PI3K persistently and class III PI3K transiently. Induces autophagy under nutrient-rich conditions and inhibits starvation-induced autophagy due to differential effects on class I versus class III PI3 kinase. Shows some limited Vps34 preference in vitro compared to PtdIns3Kgamma. However, it is typically employed in cellular studies at a concentration of 10 mM, which can inhibit all PtdIns3Ks. Can target other kinases and affect other cellular processes, such as glycogen metabolism, lysosomal acidification, endocytosis and mitochondrial permeability transition. Anticancer compound. Neuroprotective. PKA-activation dependent lipolytic agent. Enhances ATGL-dependent hydrolysis of triacylglycerols.
Catalog No. 50-205-3967
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.