Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylaminomethyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. M28695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28695G 5 g
M28691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28695G Supplier TCI America Supplier No. M28695G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90389-96-1
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD09731650
InChI Key KBRCDVSFCMVQSN-UHFFFAOYSA-N
Synonym 3-methylamino methyl benzonitrile, 3-methylaminomethyl benzonitrile, 3-methylaminomethyl-benzonitrile, benzonitrile, 3-methylamino methyl, 3-methylamino methyl-benzonitrile, 3-cyano-n-methylbenzylamine, n-methyl-n-3-cyanobenzyl amine
PubChem CID 457595
IUPAC Name 3-[(methylamino)methyl]benzonitrile
SMILES CNCC1=CC=CC(=C1)C#N

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 g
UN Number 2922
Formula Weight 146.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Methylaminomethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.