Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzo[f]quinoline, 97%, Thermo Scientific™

Catalog No. AAB2492006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2492006 5 g
AAB2492014 25 g
AAB2492022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2492006 Supplier Thermo Scientific Chemicals Supplier No. B2492006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Methylbenzo[f]quinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-06-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00046906
InChI Key SUHRSZJZYUCLOD-UHFFFAOYSA-N
Synonym 3-methylbenzo f quinoline, benzo f quinoline, 3-methyl, beta-naphthoquinaldine, .beta.-naphthoquinaldine, unii-j67ocr12xn, j67ocr12xn
PubChem CID 6797
IUPAC Name 3-methylbenzo[f]quinoline
SMILES CC1=NC2=C(C=C1)C3=CC=CC=C3C=C2

Specifications

Melting Point 73°C to 78°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material 3-Methylbenzo[f]quinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-584-1
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.