Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzofuran 98.0+%, TCI America™
SDP

Catalog No. M21121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21121G 1 g
M21125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21121G Supplier TCI America Supplier No. M21121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21535-97-7
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00205641
InChI Key ZRXHLJNBNWVNIM-UHFFFAOYSA-N
Synonym 3-methylbenzofuran, 3-methyl-benzofuran, benzofuran, 3-methyl, 3-methylbenzo b furan, acmc-209flj, #
PubChem CID 88939
IUPAC Name 3-methyl-1-benzofuran
SMILES CC1=COC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 197°C
Quantity 1 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylbenzofuran
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.