Learn More
Sigma Organic Chemistry 3-Methylbenzyl hydrazin | 1G | MFCD07785901
Supplier: Sigma Organic Chemistry CBR008271G

3-Methylbenzyl hydrazin , 1G
About this Item:
MDL #: MFCD07785901
Molecular Weight: 172.66
UNSPSC Code: 12352200
Chemical Formula: C8H13ClN2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.