Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbutanol, Solstice™

Catalog No. 60090057 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
200 L
Packaging:
Returnable Stainless Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-057 200 L Returnable Stainless Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-057 Supplier Solstice Supplier No. 59090200L
Add to Cart
Add to Cart
Request a Quote

Puriss. p.a., ACS Reagent, ≥98.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
PubChem CID 31260
ChEBI CHEBI:15837

Specifications

Boiling Point 131°C to 132°C
Flash Point 43 °C
Packaging Returnable Stainless Steel Drum
Quantity 200 L
Percent Purity min. 98.5 %
Grade ACS General Use Solvents
Physical Form Liquid
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.