Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylcinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. M129825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M129825G 25 g
M12985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M129825G Supplier TCI America Supplier No. M129825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3029-79-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00016844
InChI Key JZINNAKNHHQBOS-AATRIKPKSA-N
Synonym 3-methylcinnamic acid, m-methylcinnamic acid, e-3-m-tolyl acrylic acid, 3-m-tolyl acrylic acid, 2e-3-3-methylphenyl acrylic acid, 2e-3-3-methylphenyl prop-2-enoic acid, cinnamic acid, m-methyl, 3-methylcinnamic acid, predominantly trans, 2e-3-3-methylphenyl-2-propenoic acid, 2-propenoic acid, 3-3-methylphenyl-, 2e
PubChem CID 2063448
ChEBI CHEBI:61118
IUPAC Name (E)-3-(3-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=CC(=C1)C=CC(=O)O

Specifications

Melting Point 116°C
Color White
Quantity 25 g
Formula Weight 162.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylcinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.