Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylcyclohexylamine (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. M210525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M210525G 25 g
M21055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M210525G Supplier TCI America Supplier No. M210525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6850-35-7
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00001494
InChI Key JYDYHSHPBDZRPU-UHFFFAOYNA-N
Synonym 1-Amino-3-methylcyclohexane
PubChem CID 110931
IUPAC Name 3-methylcyclohexan-1-amine
SMILES CC1CCCC(N)C1

Specifications

Color Colorless
Boiling Point 151°C
Quantity 25 g
UN Number 2733
Formula Weight 113.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methylcyclohexylamine (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.