Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylcytosine (3-mC) Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501994143
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4143 100 μL
50-199-4144 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4143 Supplier Active Motif Supplier No. 61179
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

3-methylcytosine arises by erroneous methylation of DNA through spontaneous exposure to endogenous S-adenosyl methionine (SAM). Since the N3-position in cytosine is protected in double stranded DNA, this phenomenon mainly occurs in single stranded DNA. The resulting 3-mC is toxic and mutagenic and increased levels of 3-mC seem to impair cell proliferation. Therefore, it has to be repaired by the cell. This is established by oxidative demethylation, which is catalyzed by the AlkB protein.

Specifications

Antigen 3-Methylcytosine (3-mC)
Applications Dot Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene Alias 3mc; 3-mC; 3-meC; 3-Methyl-cytosine; C5H7NO3
Host Species Rabbit
Immunogen This 3-methylcytosine antibody was raised against 3-methylcytidine conjugated to kLH and recognizes 3-methylcytosine .
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.