Learn More
3-Methylcytosine (3-mC) Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | 3-Methylcytosine (3-mC) |
| Applications | Dot Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 30% glycerol and 0.035% sodium azide; pH |
| Gene Alias | 3mc; 3-mC; 3-meC; 3-Methyl-cytosine; C5H7NO3 |
| Host Species | Rabbit |
| Immunogen | This 3-methylcytosine antibody was raised against 3-methylcytidine conjugated to kLH and recognizes 3-methylcytosine . |
| Quantity | 10 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.