Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylpyridazine 97.0+%, TCI America™
SDP

Catalog No. M087225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M087225ML 25 mL
M08725ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M087225ML Supplier TCI America Supplier No. M087225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1632-76-4
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006469
InChI Key MXDRPNGTQDRKQM-UHFFFAOYSA-N
Synonym pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j
PubChem CID 74208
IUPAC Name 3-methylpyridazine
SMILES CC1=NN=CC=C1

Specifications

Boiling Point 215°C
Quantity 25 mL
Formula Weight 94.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylpyridazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.