Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylthio)-1-hexanol 98.0+%, TCI America™
SDP

Catalog No. M205025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M205025G 25 g
M20505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M205025G Supplier TCI America Supplier No. M205025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51755-66-9
Molecular Formula C7H16OS
Molecular Weight (g/mol) 148.26
MDL Number MFCD00010674
InChI Key JSASXSHMJYRPCM-UHFFFAOYNA-N
Synonym 3-methylthio-1-hexanol, 3-methylthio hexanol, 1-hexanol, 3-methylthio, 3-methylthio hexan-1-ol, 3-methylthiohexanol, 3-methylsulfanyl hexan-1-ol, r-3-methylthio-1-hexanol, 3-methylmercapto-1-hexanol, fema no. 3438
PubChem CID 65413
IUPAC Name 3-(methylsulfanyl)hexan-1-ol
SMILES CCCC(CCO)SC

Specifications

Color Yellow
Boiling Point 62°C
Quantity 25 g
Formula Weight 148.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)-1-hexanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.