Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylthio)propionyl Chloride 98.0+%, TCI America™
SDP

Catalog No. M080625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M080625ML 25 mL
M0806500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M080625ML Supplier TCI America Supplier No. M080625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7031-23-4
Molecular Formula C4H7ClOS
Molecular Weight (g/mol) 138.609
MDL Number MFCD00059482
InChI Key NRHMXMBVLXSSAX-UHFFFAOYSA-N
Synonym 3-(Methylmercapto)propionyl Chloride
PubChem CID 12921440
IUPAC Name 3-methylsulfanylpropanoyl chloride
SMILES CSCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 mL
UN Number 3265
Formula Weight 138.61
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)propionyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.