Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylthio)propyl Isothiocyanate 97.0+%, TCI America™
SDP

Catalog No. M202725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M202725G 25 g
M20275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M202725G Supplier TCI America Supplier No. M202725G

Chemical Identifiers

CAS 505-79-3
Molecular Formula C5H9NS2
Molecular Weight (g/mol) 147.254
MDL Number MFCD00209521
InChI Key LDKSCZJUIURGMW-UHFFFAOYSA-N
Synonym 3-methylthio propyl isothiocyanate, iberverin, 3-methylthiopropyl isothiocyanate, ibervirin, propane, 1-isothiocyanato-3-methylthio, unii-ytt13bn94z, 3-methylmercaptopropyl isothiocyanate, isothiocyanic acid, 3-methylthio propyl ester, 1-isothiocyanato-3-methylthio propane, fema no. 3312
PubChem CID 62351
IUPAC Name 1-isothiocyanato-3-methylsulfanylpropane
SMILES CSCCCN=C=S

Specifications

Color Yellow
Boiling Point 122°C
Quantity 25 g
Formula Weight 147.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)propyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.