Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylthiophene-2-carbonitrile, 96%

Catalog No. AAA1330506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1330506 5 g
AAA1330503 1 g
AAA1330514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1330506 Supplier Thermo Scientific Chemicals Supplier No. A1330506
Add to Cart

CAS: 55406-13-8 | C6H5NS | 123.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55406-13-8
Molecular Formula C6H5NS
Molecular Weight (g/mol) 123.173
MDL Number MFCD00052086
InChI Key ALZHYEITUZEZMT-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarbonitrile, 2-cyano-3-methylthiophene, acmc-1axot, 3-methyl-thiophene-2-carbonitrile, 3-methyl-2-thiophenecarbonitrile #
PubChem CID 521617
IUPAC Name 3-methylthiophene-2-carbonitrile
SMILES CC1=C(SC=C1)C#N

Specifications

Boiling Point 91°C to 93°C (12 mmHg)
Refractive Index 1.553
Quantity 5 g
UN Number UN3276
Beilstein 5495606
Formula Weight 123.18
Percent Purity 96%
Chemical Name or Material 3-Methylthiophene-2-carbonitrile
Hazard Category H301-H312+H332
Hazard Statement GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.