Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-morpholin-4-ylaniline, 97%, Thermo Scientific™

Catalog No. CC33814DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC33814DA 1 g Amber Glass Bottle
CC33814CB 250 mg Amber Glass Bottle
CC33814DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC33814DA Supplier Thermo Scientific Chemicals Supplier No. CC33814DA
Add to Cart

Chemical Identifiers

CAS 159724-40-0
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD03197165
InChI Key ZJWLMZURLIHVHE-UHFFFAOYSA-N
Synonym 3-4-morpholinyl aniline, 3-morpholinoaniline, 3-morpholinobenzenamine, 3-morpholin-4-yl aniline, benzenamine, 3-4-morpholinyl, 3-morpholin-4-yl-phenylamine, 4-3-aminophenyl morpholine, 3-morpholin-4-ylphenylamine, 3-morpholinylaniline, meta-morpholino aniline
PubChem CID 847768
IUPAC Name 3-morpholin-4-ylaniline
SMILES NC1=CC=CC(=C1)N1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 3-Morpholin-4-ylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.