Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Morpholin-4-ylmethyl)benzonitrile, 97%, Thermo Scientific™

Catalog No. CC40116DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC40116DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC40116DA Supplier Thermo Scientific Chemicals Supplier No. CC40116DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 857283-91-1
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD08060511
InChI Key GSVNKQLSALKJHW-UHFFFAOYSA-N
Synonym 3-morpholin-4-ylmethyl benzonitrile, benzonitrile,3-4-morpholinylmethyl, 3-morpholin-4-yl methyl benzonitrile, 3-morpholinomethyl benzonitrile, 3-morpholin-4-ylmethyl benzenecarbonitrile
PubChem CID 7164605
IUPAC Name 3-(morpholin-4-ylmethyl)benzonitrile
SMILES C1COCCN1CC2=CC=CC(=C2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material 3-(Morpholin-4-ylmethyl)benzonitrile
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.