Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Morpholinobenzoic acid, 97%, Thermo Scientific™

Catalog No. CC33801DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC33801DA 1 g
CC33801CB 250 mg
CC33801DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC33801DA Supplier Thermo Scientific Chemicals Supplier No. CC33801DA
Add to Cart

Chemical Identifiers

CAS 215309-00-5
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
MDL Number MFCD06659078
InChI Key VSKFQESEPGOWBS-UHFFFAOYSA-N
Synonym 3-morpholinobenzoic acid, 3-4-morpholinyl benzoic acid, 3-morpholin-4-yl-benzoic acid, 3-morpholin-4-yl benzoic acid, 3-morpholinobenzoicacid, benzoic acid, 3-4-morpholinyl, 4-3-carboxyphenyl morpholine, pubchem19534, 3-4-morpholino benzoic acid
PubChem CID 2795549
IUPAC Name 3-morpholin-4-ylbenzoic acid
SMILES C1COCCN1C2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 207.23
Percent Purity 97%
Chemical Name or Material 3-Morpholinobenzoic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.