Learn More
3-n-Octylthiophene, 97%
Description
3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).
Solubility
Soluble in organic solvents.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Color | Colorless |
| Density | 0.92 g/mL |
| Boiling Point | 106°C to 107°C (3 mmHg) |
| Flash Point | 113°C (235°F) |
| Refractive Index | 1.492 |
| Quantity | 1 g |
| Solubility Information | Soluble in organic solvents. |
| Formula Weight | 196.35 |
| Percent Purity | 97% |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H311-H332-H315-H319-H335 Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.