Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-n-Propylidenephthalide, (E)+(Z), 96%, Thermo Scientific™

Catalog No. AAA1331906
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1331906 5 g
AAA1331922 100 g
AAA1331936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1331906 Supplier Thermo Scientific Chemicals Supplier No. A1331906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17369-59-4
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00051813,MFCD00051813
InChI Key NGSZDVVHIGAMOJ-YHYXMXQVSA-N
Synonym 3-propylidenephthalide, 3-propylidene-1 3h-isobenzofuranone, unii-0vb6659n9s, propylidenephthalide, propylidene phthalide, 3e-3-propylidene-2-benzofuran-1-one, 3-propylidene phthalide, phthalide, 3-propylidene 6ci,8ci, 3e-3-propylidene-2-benzofuran-1 3h-one, fema no. 2952
PubChem CID 6259976
SMILES CC\C=C1/OC(=O)C2=CC=CC=C12

Specifications

Density 1.129
Boiling Point 101°C to 103°C (0.01 mmHg)
Flash Point 144°C (291°F)
Refractive Index 1.587
Quantity 5 g
Beilstein 4486
IUPAC Name (3Z)-3-propylidene-1,3-dihydro-2-benzofuran-1-one
Formula Weight 174.2
Percent Purity 96%
Chemical Name or Material 3-n-Propylidenephthalide, (E)+(Z)
Hazard Category H302-H317
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 241-402-8
RTECSNumber NP7235000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.