Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzenesulfonic Acid Sodium Salt Pract., 93%, Thermo Scientific™

Catalog No. AC415835000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC415835000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC415835000 Supplier Thermo Scientific Chemicals Supplier No. 415835000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-68-4
Molecular Formula C6H4NNaO5S
Molecular Weight (g/mol) 225.15
InChI Key LJRGBERXYNQPJI-UHFFFAOYSA-M
Synonym sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6
PubChem CID 31389
IUPAC Name sodium;3-nitrobenzenesulfonate
SMILES C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]

Specifications

CAS Min % 91.0
CAS Max % 100.0
Color Yellow
Loss on Drying 10% max. (135°C)
Infrared Spectrum Authentic
Linear Formula SO3NaC6H4NO2
Quantity 500 g
Beilstein 11, 68
Formula Weight 225.16
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzenesulfonic acid sodium salt, pract., 93%
Show More Show Less
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.