Learn More
3-Nitrobenzoic acid, 99%
Description
3-Nitrobenzoic acid was used to investigated the role of ozone as additional decomposition or finishing reagent in the degradation of o-, m- and p-nitobenzoic acids
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Nitrobenzoic acid was used to investigated the role of ozone as additional decomposition or finishing reagent in the degradation of o-, m- and p-nitobenzoic acids
Solubility
Soluble in water. Insoluble in acetic acid.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 140°C to 143°C |
| Density | 1.494 |
| Quantity | 5000 g |
| Beilstein | 908644 |
| Merck Index | 14,6589 |
| Solubility Information | Soluble in water. Insoluble in acetic acid. |
| Formula Weight | 167.12 |
| Percent Purity | 99% |
| Chemical Name or Material | 3-Nitrobenzoic acid |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H341-H315-H319-H335 Toxic if swallowed. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 204-508-5 |
| RTECSNumber | DH5000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.