Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. N056325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0563-25G 25 g
N05635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0563-25G Supplier TCI America Supplier No. N056325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13331-27-6
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00007193
InChI Key ZNRGSYUVFVNSAW-UHFFFAOYSA-N
PubChem CID 1677
IUPAC Name (3-nitrophenyl)boronic acid
SMILES OB(O)C1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 268°C
Color White-Yellow
Quantity 25 g
Synonym 3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid
Formula Weight 166.93
Physical Form Crystalline Powder
RTECSNumber CY8980000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.