Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrophthalic Hydrazide 97.0+%, TCI America™
SDP

Catalog No. N062225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N062225G 25 g
N06225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N062225G Supplier TCI America Supplier No. N062225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3682-15-3
Molecular Formula C8H5N3O4
Molecular Weight (g/mol) 207.15
MDL Number MFCD00006889
InChI Key YVOBBFQJMOGQOU-UHFFFAOYSA-N
Synonym 2,3-Dihydro-5-nitro-1,4-phthalazinedione, 3-Nitrophthalhydrazide
PubChem CID 3525223
IUPAC Name 5-nitro-1,2,3,4-tetrahydrophthalazine-1,4-dione
SMILES [O-][N+](=O)C1=C2C(=O)NNC(=O)C2=CC=C1

Specifications

Quantity 25 g
Formula Weight 207.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophthalic Hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.