Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Nitropropiophenone, 97%, Thermo Scientific™

Catalog No. AAA1455406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1455406 5 g
AAA1455414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1455406 Supplier Thermo Scientific Chemicals Supplier No. A1455406

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17408-16-1
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00051516
InChI Key VSPOTMOYDHRALZ-UHFFFAOYSA-N
Synonym 3'-nitropropiophenone, 1-3-nitrophenyl propan-1-one, m-nitropropiophenone, 3-nitrophenyl ethyl ketone, 1-propanone, 1-3-nitrophenyl, m-nitropropio-phenone, 3;-nitropropiophenone, acmc-209e7u, 3-nitropropiophenone, 1-3-nitrophenyl propan-l-one
PubChem CID 87096
IUPAC Name 1-(3-nitrophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 99°C to 102°C
Quantity 5 g
Beilstein 1954069
Formula Weight 179.18
Percent Purity 97%
Chemical Name or Material 3'-Nitropropiophenone
EINECSNumber 241-435-8
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.