Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Nitropropiophenone 98.0+%, TCI America™
SDP

Catalog No. N078925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N078925G 25 g
N07895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N078925G Supplier TCI America Supplier No. N078925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17408-16-1
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00051516
InChI Key VSPOTMOYDHRALZ-UHFFFAOYSA-N
Synonym 3'-nitropropiophenone, 1-3-nitrophenyl propan-1-one, m-nitropropiophenone, 3-nitrophenyl ethyl ketone, 1-propanone, 1-3-nitrophenyl, m-nitropropio-phenone, 3;-nitropropiophenone, acmc-209e7u, 3-nitropropiophenone, 1-3-nitrophenyl propan-l-one
PubChem CID 87096
IUPAC Name 1-(3-nitrophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 100°C
Color Yellow
Quantity 25 g
Formula Weight 179.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Nitropropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.