Learn More
Abcam 3-Nitrotyrosine ELISA Kit - Extracellular, 1 x 96 Tests
Supplier:  Abcam AB3152641X96TESTS
Encompass_Preferred
3-Nitrotyrosine ELISA Kit - Extracellular is a single-wash 90-min Simplestep used to quantify 3-Nitrotyrosine with a sensitivity of 41 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Competitive ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-8397
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.