Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nonen-2-one, 96%

Catalog No. AAA1908418
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1908418 50 g
AAA1908409 10 g
AAA1908430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1908418 Supplier Thermo Scientific Chemicals Supplier No. A1908418
Add to Cart

CAS: 14309-57-0 | C9H16O | 140.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14309-57-0
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00010241
InChI Key HDKLIZDXVUCLHQ-BQYQJAHWSA-N
Synonym 3-nonen-2-one, trans-3-nonen-2-one, e-non-3-en-2-one, e-3-nonen-2-one, 3-nonen-2-one, 3e, 3e-3-nonen-2-one, 3e-non-3-en-2-one, unii-fr0k39of16, 3e-nonen-2-one, 3-nonen-2-one fhfi
PubChem CID 5317045
SMILES CCCCC\C=C\C(C)=O

Specifications

Density 0.848
Boiling Point 85°C (12 mmHg)
Flash Point 81°C (177°F)
Refractive Index 1.449
Quantity 50 g
Beilstein 1743558
IUPAC Name (E)-non-3-en-2-one
Formula Weight 140.23
Percent Purity 96%
Chemical Name or Material 3-Nonen-2-one
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 238-248-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.