Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Octadecylthiophene 98.0+%, TCI America™
SDP

Catalog No. O02451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02451G 1 g
O02455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02451G Supplier TCI America Supplier No. O02451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 104934-54-5
Molecular Formula C22H40S
Molecular Weight (g/mol) 336.622
MDL Number MFCD00191567
InChI Key ARFJPHXJBIEWSZ-UHFFFAOYSA-N
Synonym 3-Stearylthiophene
PubChem CID 566857
IUPAC Name 3-octadecylthiophene
SMILES CCCCCCCCCCCCCCCCCCC1=CSC=C1

Specifications

Melting Point 36°C
Color White
Quantity 1 g
Formula Weight 336.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Octadecylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.