Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Oxo-5beta-cholanoic Acid 95.0+%, TCI America™
SDP

Catalog No. O05001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O05001G 1 g
O0500250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O05001G Supplier TCI America Supplier No. O05001G

Chemical Identifiers

CAS 1553-56-6
Molecular Formula C24H38O3
Molecular Weight (g/mol) 374.565
MDL Number MFCD00046265
InChI Key KIQFUORWRVZTHT-OPTMKGCMSA-N
PubChem CID 5283906
ChEBI CHEBI:17639
IUPAC Name (4R)-4-[(5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES CC(CCC(=O)O)C1CCC2C1(CCC3C2CCC4C3(CCC(=O)C4)C)C

Specifications

Color White
Quantity 1 g
Formula Weight 374.57
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Oxo-5beta-cholanoic Acid
RTECSNumber FZ2281400
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.