Learn More
AdipoGen 3-oxo-C10-L-HSL
SDP
Supplier:  AdipoGen CDXO0059M010
Encompass_Preferred
Chemical. CAS 147795-40-2. Formula C14H23NO4. MW 269.3. Synthetic. N-3-Oxodecanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. It was used as an autoinducer of quorum signaling by Pseudomonas putida, Yersinia enterocolitica and other Gram-negative bacteria.
Catalog No. 50-256-0307
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.