Learn More
AdipoGen N-(3-Oxoundecanoyl)-L-homoserine lactone (50 mg)
SDP
Supplier:  AdipoGen CDXO0140M050
Encompass_Preferred
Chemical. CAS: 216596-70-2. Formula: C15H25NO4. MW: 283.4. Synthetic. N-(3-Oxoundecanoyl)-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. Numerous species of bacteria employ 3OC11-HSL in cell-to-cell communication. Shown to have immune suppressive activity, inhibiting murine and human leucocyte proliferation.
Catalog No. 50-205-4581
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.