Learn More
AdipoGen 3-oxo-C8-DL-HSL
SDP
Supplier:  AdipoGen CDXO0052M100
Encompass_Preferred
Chemical. CAS 106983-27-1. Formula C12H19NO4. MW 241.3. Synthetic. N-3-Oxooctanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. This compound stimulates the tra gene expression. It is an autoinducer and potent antagonist.
Catalog No. 50-256-0319
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.