Learn More
AdipoGen 3-Oxo-dodecan-2-aminocyclohex
SDP
Supplier:  AdipoGen CDXO0034M025
Encompass_Preferred
Chemical. CAS 596104-55-1. Formula C18H31NO3. MW 309.44. Synthetic. Dodecanamide, 3-oxo-N-2-oxocyclohexyl- 3-Oxo-N-2-oxocyclohexyldodecanamide may be useful in the development and study of quorum sensing synthetic autoinducer analogs. It inhibits the lasI and rhlI reporters, pyocyanin, elastase and biofilm.
Catalog No. 50-256-0340
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.