Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenoxybenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. P096025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P096025G 25 g
P0960500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P096025G Supplier TCI America Supplier No. P096025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39515-51-0
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00003353
InChI Key MRLGCTNJRREZHZ-UHFFFAOYSA-N
Synonym m-phenoxybenzaldehyde, 3-phenoxy-benzaldehyde, benzaldehyde, 3-phenoxy, 3-formyldiphenyl ether, m-phenyloxy benzaldehyde, 3-phenoxy benzaldehyde, dsstox_cid_8005, dsstox_rid_78636, dsstox_gsid_28005, 5-phenoxybenzaldehyde
PubChem CID 38284
IUPAC Name 3-phenoxybenzaldehyde
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)C=O

Specifications

Boiling Point 184°C
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenoxybenzaldehyde
EINECSNumber (3)-3470
RTECSNumber CU7560200
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.