Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylpropionaldehyde, 94.9%, MP Biomedicals™

Catalog No. ICN15619680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15619680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15619680 Supplier MP Biomedicals, Inc Supplier No. 0215619680
Add to Cart
Add to Cart

Chemical Identifiers

CAS 104-53-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
InChI Key YGCZTXZTJXYWCO-UHFFFAOYSA-N
Synonym benzenepropanal, 3-phenylpropionaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, dihydrocinnamaldehyde, benzylacetaldehyde, 3-phenyl-1-propanal, 3-phenylpropylaldehyde, 3-phenylpropyl aldehyde, propanal, phenyl
PubChem CID 7707
IUPAC Name 3-phenylpropanal
SMILES C1=CC=C(C=C1)CCC=O

Specifications

Color Colorless
Density 1.01g/mL
Assay Percent Range 95%
Quantity 100 g
Formula Weight 134.2
Percent Purity 0.949
Physical Form Liquid
Chemical Name or Material 3-Phenylpropionaldehyde
Recommended Storage Store at 2° to 8°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.