Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylpropyl Chloride 97.0+%, TCI America™
SDP

Catalog No. P1262500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1262500G 500 g
P126225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1262500G Supplier TCI America Supplier No. P1262500G

Chemical Identifiers

CAS 104-52-9
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.637
MDL Number MFCD00001001
InChI Key XZBXAYCCBFTQHH-UHFFFAOYSA-N
Synonym 3-chloropropyl benzene, 1-chloro-3-phenylpropane, 3-phenylpropyl chloride, benzene, 3-chloropropyl, 3-phenyl-1-chloropropane, 3-chloro-1-phenylpropane, .gamma.-chloropropylbenzene, benzene, chloropropyl, .gamma.-phenylpropyl chloride, 3-penylpropylchloride
PubChem CID 7706
IUPAC Name 3-chloropropylbenzene
SMILES C1=CC=C(C=C1)CCCCl

Specifications

Color Yellow
Boiling Point 219°C
Quantity 500 g
Formula Weight 154.64
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenylpropyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.