Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylpyridine 94.0+%, TCI America™
SDP

Catalog No. P104025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P104025G 25 g
P10405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P104025G Supplier TCI America Supplier No. P104025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1008-88-4
Molecular Formula C11H9N
Molecular Weight (g/mol) 155.2
MDL Number MFCD00006380
InChI Key HJKGBRPNSJADMB-UHFFFAOYSA-N
Synonym pyridine, 3-phenyl, 3-phenyl-pyridine, m-phenylpyridine, unii-edz6kb7jvc, 5-phenylpyridine, 3-phenyipyridine, edz6kb7jvc, 3-azabiphenyl, 3-pyridylbenzene, 3-phenyl pyridine
PubChem CID 13886
IUPAC Name 3-phenylpyridine
SMILES C1=CC=C(C=C1)C2=CN=CC=C2

Specifications

Boiling Point 274°C
Quantity 25 g
Formula Weight 155.20
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.