Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylsalicylic Acid 98.0+%, TCI America™
SDP

Catalog No. P022625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P022625G 25 g
P02265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P022625G Supplier TCI America Supplier No. P022625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 304-06-3
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00020257
InChI Key ZJWUEJOPKFYFQD-UHFFFAOYSA-N
Synonym 2-Hydroxybiphenyl-3-carboxylic Acid
PubChem CID 9348
IUPAC Name 2-hydroxy-3-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=C(C(=CC=C2)C(=O)O)O

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylsalicylic Acid
EINECSNumber (4)-0023
RTECSNumber VO5600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.