Learn More
Description
3'-Phosphoadenosine-5'-phosphosulfate (PAPS) is the universal sulfur donor for sulfotransferases. It is synthesized from ATP through two steps. In the first step, one ATP molecule is converted to adenosine-5'-phosphosulfate (APS) by ATP sulfurylase and pyrophosphate (PPi) is generated as a byproduct. In the second step, the APS is converted to PAPS by APS kinase and ADP is generated as a byproduct. To drive the reaction forward, the pyrophosphate can be further degraded to phosphate (Pi) by inorganic pyrophosphatase to eliminate feedback inhibition, and the ADP can be converted back to ATP using pyruvate kinase.
- Suitable for kinetic assay of sulfotransferases using the Universal Sulfotransferase Activity Kit (Mfr. No. EA003) or for enzymatic synthesis of sulfonated molecules
- Purity >90% of the nucleotide based on HPLC: No Li+ or Na+ ions, and free inorganic phosphate <0.5%
Specifications
Specifications
| Product Line | R&D Systems |
| Quantity | 500 μg |
| Product Type | PAPS, C10H15N5O13P2S |
| Storage Requirements | Store the unopened product at -70° C for up to six months. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
