Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Picolylamine 99.0+%, TCI America™
SDP

Catalog No. P0762100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0762100ML 100 mL
P076225ML 25 mL
P0762500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0762100ML Supplier TCI America Supplier No. P0762100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3731-52-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006412
InChI Key HDOUGSFASVGDCS-UHFFFAOYSA-N
Synonym 3-aminomethyl pyridine, 3-pyridinemethanamine, 3-picolylamine, picolamine, 3-pyridylmethylamine, 3-pyridinylmethanamine, 3-pyridinemethylamine, 3-aminomethylpyridine, pyridin-3-yl methanamine, 3-pyridylmethanamine
PubChem CID 31018
IUPAC Name pyridin-3-ylmethanamine
SMILES C1=CC(=CN=C1)CN

Specifications

Melting Point -21°C
Color Yellow
pH 11 to 12 (10% H2O 20°C)
Boiling Point 74°C
Quantity 100 mL
UN Number 2735
Formula Weight 108.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Picolylamine
EINECSNumber (5)-6097
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.