Learn More
Chemglass Life Sciences Vacuum Manifold, Left Handed, 3 Ports, 0-4mm Chem-Cap Valve, 240mm Overall width
SDP
Supplier:  Chemglass Life Sciences CG443407
Encompass_Preferred
Vacuum Manifold, Left Handed, 3 Ports, 0-4mm Chem-Cap Valve, 240mm Overall width. General purpose single bank vacuum manifold with 3 ports. Main tube is 38mm OD with a serrated hose connection, having the smallest serration 13.5mm and the largest serration 15.5mm, located at either the left or right side for vacuum connection. Ports are 0-4mm Chem-Vac™ Chem-Cap valves with serrated hose connections. The 0-4mm has the smallest serration 10mm and the largest serration 11mm. Ports are spaced 60mm center to center for the 0-4mm size.
Catalog No. CG443407
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.