Learn More
United Scientific™ 3-Prong Clamp With Flexible Extension Rod
SDPSCI_ED
Catalog No. S89609 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Silver
Catalog No. Color
S89609 Silver
1 options
This item is not returnable. View return policy
Catalog No. S89609 Supplier United Scientific™ Supplier No. UNFLEXCLP01
Add to Cart
Add to Cart
This item is not returnable. View return policy
This United Scientific™ 3-Prong Clamp With Flexible Extension Rod is extremely versatile and can be placed in virtually any position or angle.
This flexible arm is extremely versatile and can be placed in virtually any position or angle.
  • Includes a 3-Prong Clamp Head, nickel-plated flex arm in 305 mm (12 in.) length
  • Lab-Frame Connector securely attaches to support stands, lab-frames; or any support rod up to 19 mm (0.75 in.) diameter
Specifications
Quantity 1 Ea.
Type Flexible Clamp
Material Nickel-plated
Color Silver
Jaw Type 3-Prong
Diameter (Metric) Inside 95.3 mm